Merck
  • Knock-Down of the IFR1 Protein Perturbs the Homeostasis of Reactive Electrophile Species and Boosts Photosynthetic Hydrogen Production in
Knock-Down of the IFR1 Protein Perturbs the Homeostasis of Reactive Electrophile Species and Boosts Photosynthetic Hydrogen Production in
Frontiers in plant science (2017-08-22)
Deepak Venkanna, Christian Südfeld, Thomas Baier, Sarah V Homburg, Anant V Patel, Lutz Wobbe, Olaf Kruse
ABSTRACT
The protein superfamily of short-chain dehydrogenases/reductases (SDR), including members of the atypical type (aSDR), covers a huge range of catalyzed reactions and
Sigma-Aldrich
2,5-Dibromo-6-isopropyl-3-methyl-1,4-benzoquinone