Merck
  • Synthesis and characterization of functionalized mesoprous SBA-15 decorated with Fe(3)O(4) nanoparticles for removal of Ce(III) ions from aqueous solution: ICP-OES detection and central composite design optimization.
Synthesis and characterization of functionalized mesoprous SBA-15 decorated with Fe(3)O(4) nanoparticles for removal of Ce(III) ions from aqueous solution: ICP-OES detection and central composite design optimization.
Journal of colloid and interface science (2017-02-06)
Kheibar Dashtian, Rouholah Zare-Dorabei
ABSTRACT
A selective adsorbent based on the modification of mesoprous SBA-15 with N,N'-bis(salicylidene)-1,3-ethylenediamine Schiff base and decorated with Fe
Sigma-Aldrich
(3-Chloropropyl)triethoxysilane, 95%