Merck
  • iRhom2 regulates CSF1R cell surface expression and non-steady state myelopoiesis in mice.
iRhom2 regulates CSF1R cell surface expression and non-steady state myelopoiesis in mice.
European journal of immunology (2016-09-08)
Xiaoping Qing, Lindsay Rogers, Arthur Mortha, Yonit Lavin, Patricia Redecha, Priya D Issuree, Thorsten Maretzky, Miriam Merad, David McIlwain, Tak W Mak, Christopher M Overall, Carl P Blobel, Jane E Salmon
ABSTRACT
CSF1R (colony stimulating factor 1 receptor) is the main receptor for CSF1 and has crucial roles in regulating myelopoeisis. CSF1R can be proteolytically released from the cell surface by ADAM17 (A disintegrin and metalloprotease 17). Here, we identified CSF1R as a major substrate of ADAM17 in an unbiased degradomics screen. We explored the impact of CSF1R shedding by ADAM17 and its upstream regulator, inactive rhomboid protein 2 (iRhom2, gene name Rhbdf2), on homeostatic development of mouse myeloid cells. In iRhom2-/- mice, we found constitutive accumulation of membrane-bound CSF1R on myeloid cells at steady state, although cell numbers of these populations were not altered. However, in the context of mixed bone marrow (BM) chimera, under competitive pressure, iRhom2-/- BM progenitor-derived monocytes, tissue macrophages and lung DCs showed a repopulation advantage over those derived from wild-type (WT) BM progenitors, suggesting enhanced CSF1R signaling in the absence of iRhom2. In vitro experiments indicate that iRhom2-/- Lin
Sigma-Aldrich
Sodium cyanoborodeuteride, 97 atom % D, ≥96% (CP)