Merck
  • Evaluation of Host-Plant Resistance of Selected Rice Genotypes to the Rice Water Weevil (Coleoptera: Curculionidae).
Evaluation of Host-Plant Resistance of Selected Rice Genotypes to the Rice Water Weevil (Coleoptera: Curculionidae).
Environmental entomology (2016-12-29)
Suhas S Vyavhare, David R Gealy, Michael O Way, Rodante E Tabien, Rebecca A Pearson
ABSTRACT
The rice water weevil, Lissorhoptrus oryzophilus Kuschel, is the most important insect pest of rice in the United States. Management of L. oryzophilus mainly depends upon the use of insecticides due to the lack of effective alternative management tactics. A 3-yr field study was conducted to determine if difference exists among rice genotypes and cultivars of inbred tropical japonica subspecies commercially grown in the southern United States [Cocodrie (PI 606331), CL171, and CL151 (PI 654463)] and the germplasm lines of indica subspecies adapted to tropical climates of Asia [WC 4644 (PI 312777), TNI (PI 495830), Rondo (PI 615022), 4612 (PI 615039), TeQing (PI 536047), and 4593 (PI 615031)] for resistance to L. oryzophilus Experiments were established as a split-plot design with cultivars as main plots and insecticide treatment as subplots. No significant differences were observed in number of L. oryzophilus larvae recovered across cultivars and genotypes, indicating no significant variation in their preference to L. oryzophilus oviposition. Insecticide treatment had a significant impact on L. oryzophilus larval density. However, grain yield did not vary significantly between treated and untreated plots for any of the cultivars and genotypes. The amount of yield loss in response to L. oryzophilus infestation did not vary significantly across genotypes and cultivars, indicating no variation among these genotypes for resistance to L. oryzophilus.
Product Description
Supelco
Chlorantraniliprole, PESTANAL®, analytical standard