Merck
  • β-N-oxalyl-L-α, β- diaminopropionic acid induces HRE expression by inhibiting HIF-prolyl hydroxylase-2 in normoxic conditions.
β-N-oxalyl-L-α, β- diaminopropionic acid induces HRE expression by inhibiting HIF-prolyl hydroxylase-2 in normoxic conditions.
European journal of pharmacology (2016-10-30)
Ravi Kumar Eslavath, Deepshikha Sharma, Nabil A M Bin Omar, Rajasekhar Chikati, Mahesh Kumar Teli, G K Rajanikant, Surya S Singh
ABSTRACT
Hypoxia inducible factor (HIF)-1α, a subunit of HIF transcription factor, regulates cellular response to hypoxia. In normoxic conditions, it is hydroxylated by prolyl hydroxylase (PHD)-2 and targeted for proteosomal degradation. Drugs which inhibit PHD-2 have implications in conditions arising from insufficient blood supply. β-ODAP (β-N- oxalyl-L-α, β- diaminopropionic acid), a non-protein excitatory amino acid present in Lathyrus sativus, is an α-amino-3-hydroxy-5-methyl-4-isoxazole propionic acid receptor agonist known to activate conventional protein kinase C and stabilize HIF-1α under normoxic conditions. However, the mechanism of HIF-1α stabilization by this compound is unknown. In silico approach was used to understand the mechanism of stabilization of HIF-1α which revealed β-ODAP interacts with key amino acid residues and Fe
Sigma-Aldrich
β-ODAP, ≥98% (HPLC)