Merck
  • Toxicokinetics of titanium dioxide (TiO2) nanoparticles after inhalation in rats.
Toxicokinetics of titanium dioxide (TiO2) nanoparticles after inhalation in rats.
Toxicology letters (2016-11-26)
Igor Pujalté, Denis Dieme, Sami Haddad, Alessandra Maria Serventi, Michèle Bouchard
ABSTRACT
This study focused on the generation of aerosols of titanium dioxide (TiO2) nanoparticles (NPs) and their disposition kinetics in rats. Male Sprague-Dawley rats were exposed by inhalation to 15mg/m3 of anatase TiO2 NPs (∼20nm) during 6h. Rats were sacrificed at different time points over 14days following the onset of inhalation. Ti levels were quantified by ICP-MS in blood, tissues, and excreta. Oxidative damages were also monitored (MDA). Highest tissue levels of Ti were found in lungs; peak values were reached only at 48h followed by a progressive decrease over 14days, suggesting a persistence of NPs at the site-of-entry. Levels reached in blood, lymph nodes and other internal organs (including liver, kidney, spleen) were circa one order of magnitude lower than in lungs, but the profiles were indicative of a certain translocation to the systemic circulation. Large amounts were recovered in feces compared to urine, suggesting that inhaled NPs were eliminated mainly by mucociliary clearance and ingested. TiO2 NPs also appeared to be partly transferred to olfactory bulbs and brain. MDA levels indicative of oxidative damage were significantly increased in lungs and blood at 24h but this was not clearly reflected at later times. Translocation and clearance rates of inhaled NPs under different realistic exposure conditions should be further documented.
Product Description
Supelco
Indium Standard for ICP, TraceCERT®, 1 g/L In in nitric acid (nominal concentration)