Merck
  • Transcriptional dysregulation causes altered modulation of inhibition by haloperidol.
Transcriptional dysregulation causes altered modulation of inhibition by haloperidol.
Neuropharmacology (2016-08-03)
Lillian J Brady, Aundrea F Bartley, Qin Li, Laura J McMeekin, John J Hablitz, Rita M Cowell, Lynn E Dobrunz
ABSTRACT
Many neuropsychiatric and neurodevelopmental disorders such as schizophrenia and autism involve interneuron transcriptional dysregulation. The transcriptional coactivator PGC-1α regulates gene expression in GABAergic interneurons, which are important for regulating hippocampal network activity. Genetic deletion of PGC-1α causes a decrease in parvalbumin expression, similar to what is observed in schizophrenia postmortem tissue. Our lab has previously shown that PGC-1α
Sigma-Aldrich
L-741,626, ≥98% (HPLC)