Merck
  • Cdk phosphorylation triggers sequential intramolecular interactions that progressively block Rb functions as cells move through G1.
Cdk phosphorylation triggers sequential intramolecular interactions that progressively block Rb functions as cells move through G1.
Cell (1999-09-28)
J W Harbour, R X Luo, A Dei Santi, A A Postigo, D C Dean
ABSTRACT
We present evidence that phosphorylation of the C-terminal region of Rb by Cdk4/6 initiates successive intramolecular interactions between the C-terminal region and the central pocket. The initial interaction displaces histone deacetylase from the pocket, blocking active transcriptional repression by Rb. This facilitates a second interaction that leads to phosphorylation of the pocket by Cdk2 and disruption of pocket structure. These intramolecular interactions provide a molecular basis for sequential phosphorylation of Rb by Cdk4/6 and Cdk2. Cdk4/6 is activated early in G1, blocking active repression by Rb. However, it is not until near the end of G1, when cyclin E is expressed and Cdk2 is activated, that Rb is prevented from binding and inactivating E2F.
Sigma-Aldrich
CDK4/CyclinD3, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
CDK4/Cyclin D1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution