Merck
Synthesis of fluorinated 1-(3-morpholin-4-yl-phenyl)-ethylamines.
Bioorganic & medicinal chemistry letters (2003-05-06)
Yong-Jin Wu, Huan He, Li-Qiang Sun, Dedong Wu, Qi Gao, Hui-Yin Li
ABSTRACT
The synthesis of four (+/-)-fluorinated 1-(3-morpholin-4-yl-phenyl)-ethylamines and an enantioselective approach to these amines through reductive amination are described.