Merck
  • Human alpha-synuclein-harboring familial Parkinson's disease-linked Ala-53 --> Thr mutation causes neurodegenerative disease with alpha-synuclein aggregation in transgenic mice.
Human alpha-synuclein-harboring familial Parkinson's disease-linked Ala-53 --> Thr mutation causes neurodegenerative disease with alpha-synuclein aggregation in transgenic mice.
Proceedings of the National Academy of Sciences of the United States of America (2002-06-27)
Michael K Lee, Wanda Stirling, Yanqun Xu, Xueying Xu, Dike Qui, Allen S Mandir, Ted M Dawson, Neal G Copeland, Nancy A Jenkins, Don L Price
ABSTRACT
Mutations in alpha-synuclein (alpha-Syn) cause Parkinson's disease (PD) in a small number of pedigrees with familial PD. Moreover, alpha-Syn accumulates as a major component of Lewy bodies and Lewy neurites, intraneuronal inclusions that are neuropathological hallmarks of PD. To better understand the pathogenic relationship between alterations in the biology of alpha-Syn and PD-associated neurodegeneration, we generated multiple lines of transgenic mice expressing high levels of either wild-type or familial PD-linked Ala-30 --> Pro (A30P) or Ala-53 --> Thr (A53T) human alpha-Syns. The mice expressing the A53T human alpha-Syn, but not wild-type or the A30P variants, develop adult-onset neurodegenerative disease with a progressive motoric dysfunction leading to death. Pathologically, affected mice exhibit neuronal abnormalities (in perikarya and neurites) including pathological accumulations of alpha-Syn and ubiquitin. Consistent with abnormal neuronal accumulation of alpha-Syn, brain regions with pathology exhibit increases in detergent-insoluble alpha-Syn and alpha-Syn aggregates. Our results demonstrate that the A53T mutant alpha-Syn causes significantly greater in vivo neurotoxicity as compared with other alpha-Syn variants. Further, alpha-Syn-dependent neurodegeneration is associated with abnormal accumulation of detergent-insoluble alpha-Syn.
Sigma-Aldrich
Anti-Synuclein β Antibody, serum, Chemicon®
Sigma-Aldrich
α-Synuclein A53T human, recombinant, expressed in E. coli, N-terminal histidine tagged, ≥90% (SDS-PAGE), lyophilized powder