Merck
  • Molecular analysis of the subtype-selective inhibition of cloned KATP channels by PNU-37883A.
Molecular analysis of the subtype-selective inhibition of cloned KATP channels by PNU-37883A.
British journal of pharmacology (2004-02-06)
H Kovalev, J M Quayle, T Kamishima, D Lodwick
ABSTRACT
1. In this study, we have used Kir6.1/Kir6.2 chimeric proteins and current recordings to investigate the molecular basis of PNU-37883A inhibition of cloned K(ATP) channels. 2. Rat Kir6.1, Kir6.2 and Kir6.1/Kir6.2 chimeras were co-expressed with either SUR2B or SUR1, following RNA injection into Xenopus oocytes, and fractional inhibition of K(ATP) currents by 10 microm PNU-37883A reported. 3. Channels containing Kir6.1/SUR2B were more sensitive to inhibition by PNU-37883A than those containing Kir6.2/SUR2B (mean fractional inhibition: 0.70, cf. 0.07). 4. On expression with SUR2B, a chimeric channel with the Kir6.1 pore and the Kir6.2 amino- and carboxy-terminal domains was PNU-37883A insensitive (0.06). A chimera with the Kir6.1 carboxy-terminus and Kir6.2 amino-terminus and pore was inhibited (0.48). These results, and those obtained with other chimeras, suggest that the C-terminus is an important determinant of PNU-37883A inhibition of Kir6.1. Similar results were seen when constructs were co-expressed with SUR1. Further chimeric constructs localised PNU-37883A sensitivity to an 81 amino-acid residue section in the Kir6.1 carboxy-terminus. 5. Our data show that structural differences between Kir6.1 and Kir6.2 are important in determining sensitivity to PNU-37883A. This compound may prove useful in probing the structural and functional differences between the two channel subtypes.