Merck
  • Human c-myc onc gene is located on the region of chromosome 8 that is translocated in Burkitt lymphoma cells.
Human c-myc onc gene is located on the region of chromosome 8 that is translocated in Burkitt lymphoma cells.
Proceedings of the National Academy of Sciences of the United States of America (1982-12-01)
R Dalla-Favera, M Bregni, J Erikson, D Patterson, R C Gallo, C M Croce
ABSTRACT
Human sequences related to the transforming gene (v-myc) of avian myelocytomatosis virus (MC29) are represented by at least one gene and several related sequences that may represent pseudogenes. By using a DNA probe that is specific for the complete gene (c-myc), different somatic cell hybrids possessing varying numbers of human chromosomes were analyzed by the Southern blotting technique. The results indicate that the human c-myc gene is located on chromosome 8. The analysis of hybrids between rodent cells and human Burkitt lymphoma cells, which carry a reciprocal translocation between chromosomes 8 and 14, allowed the mapping of the human c-myc gene on region (q24 leads to qter) of chromosome 8. This chromosomal region is translocated to either human chromosome 2, 14, or 22 in Burkitt lymphoma cells.
Product Description
Roche
Anti-c-myc, from mouse IgG1κ