- Anticancer effects of CKD-602 (Camtobell
Anticancer effects of CKD-602 (Camtobell
CKD-602 (7-[2-(N-isopropylamino) ethyl]-(20S)-camptothecin, belotecan) is a synthetic water-soluble camptothecin derivative and topoisomerase I inhibitor that has been shown to exert a clinical anticancer effect on various types of tumor. In the present study, the anticancer effects of CKD-602 on the following three human oral squamous cell carcinoma (OSCC) cell lines originating from Korean cancer patients: YD-8 (tongue), YD-9 (buccal mucosa) and YD-38 (lower gingiva) were analyzed. The apoptotic proportion of the cells and cell cycle position were analyzed using flow cytometry. The expression of cell cycle regulatory proteins was detected by western blot analysis. CKD-602 was demonstrated to exert a time- and dose-dependent antiproliferative effect in all cell lines