Merck
  • Dendritic cell-bound IgE functions to restrain allergic inflammation at mucosal sites.
Dendritic cell-bound IgE functions to restrain allergic inflammation at mucosal sites.
Mucosal immunology (2014-09-18)
B Platzer, K Baker, M P Vera, K Singer, M Panduro, W S Lexmond, D Turner, S O Vargas, J-P Kinet, D Maurer, R M Baron, R S Blumberg, E Fiebiger
ABSTRACT
Antigen-mediated cross-linking of Immunoglobulin E (IgE) bound to mast cells/basophils via FcɛRI, the high affinity IgE Fc-receptor, is a well-known trigger of allergy. In humans, but not mice, dendritic cells (DCs) also express FcɛRI that is constitutively occupied with IgE. In contrast to mast cells/basophils, the consequences of IgE/FcɛRI signals for DC function remain poorly understood. We show that humanized mice that express FcɛRI on DCs carry IgE like non-allergic humans and do not develop spontaneous allergies. Antigen-specific IgE/FcɛRI cross-linking fails to induce maturation or production of inflammatory mediators in human DCs and FcɛRI-humanized DCs. Furthermore, conferring expression of FcɛRI to DCs decreases the severity of food allergy and asthma in disease-relevant models suggesting anti-inflammatory IgE/FcɛRI signals. Consistent with the improved clinical parameters in vivo, antigen-specific IgE/FcɛRI cross-linking on papain or lipopolysaccharide-stimulated DCs inhibits the production of pro-inflammatory cytokines and chemokines. Migration assays confirm that the IgE-dependent decrease in cytokine production results in diminished recruitment of mast cell progenitors; providing a mechanistic explanation for the reduced mast cell-dependent allergic phenotype observed in FcɛRI-humanized mice. Our study demonstrates a novel immune regulatory function of IgE and proposes that DC-intrinsic IgE signals serve as a feedback mechanism to restrain allergic tissue inflammation.
Sigma-Aldrich
Interleukin-4 human, IL-4, potency:, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
IL-4 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Interleukin-4 from rat, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥97% (SDS-PAGE)
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
IL-4 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-4 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Interleukin-4 from mouse, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-4 human, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture