Merck
  • GMDP: unusual physico-chemical and biological properties of the anomeriс forms.
GMDP: unusual physico-chemical and biological properties of the anomeriс forms.
Journal of peptide science : an official publication of the European Peptide Society (2015-07-15)
Elena A Meshcheryakova, Konstantin S Mineev, Pavel E Volynski, Tatiana M Andronova, Vadim T Ivanov
ABSTRACT
Disaccharide containing unit of peptidoglycan from bacterial cell wall, N-acetyl-d-glucosaminyl-N-acetylmuramyl-l-alanyl-d-glutaminamide (gluсosaminyl-muramyl-dipeptide) registered in Russia as an immunomodulatory drug, is shown to participate in slow equilibrium of α and β anomeric forms. Data of NMR spectra and molecular dynamics indicate that the α-anomer predominantly acquires a folded conformation stabilized by intramolecular hydrogen bond between the alanyl carbonyl and muramyl NH proton. The β-form displays a considerable fraction of extended, non-hydrogen bonded structures. In the standard immunoadjuvant test system, the α-form is practically inactive, and the activity of the equilibrium mixture with α : β = 68 : 32 ratio is due to the presence of β-anomer. Such unique α-β selectivity of biological action must be considered at the design of related immunoactive glycopeptides.
Sigma-Aldrich
Muramic acid, ≥95% (TLC)