Merck
  • Small-molecule inhibition of oncogenic eukaryotic protein translation in mesothelioma cells.
Small-molecule inhibition of oncogenic eukaryotic protein translation in mesothelioma cells.
Investigational new drugs (2014-04-09)
Esther Z Chen, Blake A Jacobson, Manish R Patel, Aniekan M Okon, Shui Li, Kerry Xiong, Abhishek J Vaidya, Peter B Bitterman, Carston R Wagner, Robert A Kratzke
ABSTRACT
Deranged cap-mediated translation is implicated in the genesis, maintenance and progression of many human cancers including mesothelioma. In this study, disrupting the eIF4F complex by antagonizing the eIF4E-mRNA-cap interaction is assessed as a therapy for mesothelioma. Mesothelioma cells were treated with 4Ei-1, a membrane permeable prodrug that when converted to the active drug, 7-benzyl guanosine monophosphate (7Bn-GMP) displaces capped mRNAs from the eIF4F complex. Colony formation was measured in mesothelioma treated with 4Ei-1 alone or combined with pemetrexed. Proliferation was examined in cells treated with 4Ei-1. Binding to a synthetic cap-analogue was used to study the strength of eIF4F complex activation in lysates exposed to 4Ei-1. 4Ei-1 treatment resulted in a dose dependent decrease in colony formation and cell viability. Combination therapy of 4Ei-1 with pemetrexed further reduced colony number. Formation of eIF4F cap-complex decreased in response to 4Ei-1 exposure. 4Ei-1 is a novel prodrug that reduces proliferation, represses colony formation, diminishes association of eIF4F with the mRNA cap, and sensitizes mesothelioma cells to pemetrexed.
Sigma-Aldrich
7-Methylguanosine, ≥90%