Merck
  • Ultrasensitive electrochemical detection of secretoneurin based on Pb(2+)-decorated reduced graphene oxide-tetraethylene pentamine as a label.
Ultrasensitive electrochemical detection of secretoneurin based on Pb(2+)-decorated reduced graphene oxide-tetraethylene pentamine as a label.
Biosensors & bioelectronics (2015-02-24)
Guolin Yuan, Huali Chen, Chunyong Xia, Liuliu Gao, Chao Yu
ABSTRACT
In this work, a novel electrochemical immunosensor for the detection of secretoneurin (SN), which uses metal ion functionalised reduced graphene oxide-tetraethylene pentamine (rGO-TEPA) as a label, is reported for the first time. rGO-TEPA contains a large number of amino groups, which makes it an ideal templet for the loading of metal ions. rGO-TEPA-Pb(2+) was employed to immobilise secondary secretoneurin (SN) antibody (Ab2), and the resulting nanocomposite (Ab2-rGO-TEPA-Pb(2+)) was used as a trace tag for signal amplification. A modified electrode consisting of functionalised graphene nanosheets (Au@GS) was used as a substrate to immobilise the antibodies. Under the optimal conditions, the immunoassay exhibited high sensitivity, acceptable stability and reproducibility with a wide linear range from 0.001 to 100ngmL(-1) (R=0.996), and an ultra-low detection limit of 0.33pgmL(-1) (S/N=3). Furthermore, the immunosensor could be employed to detect SN in clinical serum samples. The proposed sensing strategy enriches the electrochemical immunoassay and exhibits potential for the point-of-care diagnostic application of the clinical screening of biomarkers.
Sigma-Aldrich
Reduced graphene oxide, tetraethylene pentamine functionalized