Merck
  • Effectiveness of photocatalytic filter for removing volatile organic compounds in the heating, ventilation, and air conditioning system.
Effectiveness of photocatalytic filter for removing volatile organic compounds in the heating, ventilation, and air conditioning system.
Journal of the Air & Waste Management Association (1995) (2006-06-03)
Kuo-Pin Yu, Grace Whei-May Lee, Wei-Ming Huang, Chih-Cheng Wu, Chia-ling Lou, Shinhao Yang
ABSTRACT
Nowadays, the heating, ventilation, and air conditioning (HVAC) system has been an important facility for maintaining indoor air quality. However, the primary function of typical HVAC systems is to control the temperature and humidity of the supply air. Most indoor air pollutants, such as volatile organic compounds (VOCs), cannot be removed by typical HVAC systems. Thus, some air handling units for removing VOCs should be added in typical HVAC systems. Among all of the air cleaning techniques used to remove indoor VOCs, photocatalytic oxidation is an attractive alternative technique for indoor air purification and deodorization. The objective of this research is to investigate the VOC removal efficiency of the photocatalytic filter in a HVAC system. Toluene and formaldehyde were chosen as the target pollutants. The experiments were conducted in a stainless steel chamber equipped with a simplified HVAC system. A mechanical filter coated with Degussa P25 titania photocatalyst and two commercial photocatalytic filters were used as the photocatalytic filters in this simplified HVAC system. The total air change rates were controlled at 0.5, 0.75, 1, 1.25, and 1.5 hr(-1), and the relative humidity (RH) was controlled at 30%, 50%, and 70%. The ultraviolet lamp used was a 4-W, ultraviolet-C (central wavelength at 254 nm) strip light bulb. The first-order decay constant of toluene and formaldehyde found in this study ranged from 0.381 to 1.01 hr(-1) under different total air change rates, from 0.34 to 0.433 hr(-1) under different RH, and from 0.381 to 0.433 hr(-1) for different photocatalytic filters.
Product Description
Supelco
LpDNPH S10 Cartridge, bed wt. 350 mg, volume 3 mL, pkg of 50 ea