Merck
  • Nitrilase and Fhit homologs are encoded as fusion proteins in Drosophila melanogaster and Caenorhabditis elegans.
Nitrilase and Fhit homologs are encoded as fusion proteins in Drosophila melanogaster and Caenorhabditis elegans.
Proceedings of the National Academy of Sciences of the United States of America (1998-07-22)
Y Pekarsky, M Campiglio, Z Siprashvili, T Druck, Y Sedkov, S Tillib, A Draganescu, P Wermuth, J H Rothman, K Huebner, A M Buchberg, A Mazo, C Brenner, C M Croce
ABSTRACT
The tumor suppressor gene FHIT encompasses the common human chromosomal fragile site at 3p14.2 and numerous cancer cell biallelic deletions. To study Fhit function we cloned and characterized FHIT genes from Drosophila melanogaster and Caenorhabditis elegans. Both genes code for fusion proteins in which the Fhit domain is fused with a novel domain showing homology to bacterial and plant nitrilases; the D. melanogaster fusion protein exhibited diadenosine triphosphate (ApppA) hydrolase activity expected of an authentic Fhit homolog. In human and mouse, the nitrilase homologs and Fhit are encoded by two different genes: FHIT and NIT1, localized on chromosomes 3 and 1 in human, and 14 and 1 in mouse, respectively. We cloned and characterized human and murine NIT1 genes and determined their exon-intron structure, patterns of expression, and alternative processing of their mRNAs. The tissue specificity of expression of murine Fhit and Nit1 genes was nearly identical. Because fusion proteins with dual or triple enzymatic activities have been found to carry out specific steps in a given biochemical or biosynthetic pathway, we postulate that Fhit and Nit1 likewise collaborate in a biochemical or cellular pathway in mammalian cells.