- The polyoma virus large T binding protein p150 is a transcriptional repressor of c-MYC.
The polyoma virus large T binding protein p150 is a transcriptional repressor of c-MYC.
PloS one (2012-10-03)
Chang Kyoo Sung, Hyungshin Yim, Hongcang Gu, Dawei Li, Erik Andrews, Sekhar Duraisamy, Cheng Li, Ronny Drapkin, Thomas Benjamin
PMID23029531
ABSTRACT
p150, product of the SALL2 gene, is a binding partner of the polyoma virus large T antigen and a putative tumor suppressor. p150 binds to the nuclease hypersensitive element of the c-MYC promoter and represses c-MYC transcription. Overexpression of p150 in human ovarian surface epithelial cells leads to decreased expression, and downregulation to increased expression, of c-MYC. c-MYC is repressed upon restoration of p150 to ovarian carcinoma cells. Induction of apoptosis by etoposide results in recruitment of p150 to the c-MYC promoter and to repression of c-MYC. Analysis of data in The Cancer Genome Atlas shows negative correlations between SALL2 and c-MYC expression in four common solid tumor types.