- Arylation of diazoesters by a transient N-H insertion organocascade.
Arylation of diazoesters by a transient N-H insertion organocascade.
Angewandte Chemie (International ed. in English) (2013-09-17)
Tyler J Auvil, Sonia S So, Anita E Mattson
PMID24038873
ABSTRACT
It takes two: A unique organocatalyzed cascade for the unsymmetric double arylation of α-nitrodiazoesters is described. This organocascade features the strategic use of carbene-activating anilines in conjunction with a urea catalyst, thus allowing for the synthesis of pharmaceutically attractive α-diarylesters through a transient NH insertion process.