Merck
  • Chikungunya virus nsP3 blocks stress granule assembly by recruitment of G3BP into cytoplasmic foci.
Chikungunya virus nsP3 blocks stress granule assembly by recruitment of G3BP into cytoplasmic foci.
Journal of virology (2012-07-28)
Jelke J Fros, Natalia E Domeradzka, Jim Baggen, Corinne Geertsema, Jacky Flipse, Just M Vlak, Gorben P Pijlman
ABSTRACT
Chikungunya virus nonstructural protein nsP3 has an essential but unknown role in alphavirus replication and interacts with Ras-GAP SH3 domain-binding protein (G3BP). Here we describe the first known function of nsP3, to inhibit stress granule assembly by recruiting G3BP into cytoplasmic foci. A conserved SH3 domain-binding motif in nsP3 is essential for both nsP3-G3BP interactions and viral RNA replication. This study reveals a novel role for nsP3 as a regulator of the cellular stress response.