Merck
  • A new method of anomeric protection and activation based on the conversion of glycosyl azides into glycosyl fluorides.
A new method of anomeric protection and activation based on the conversion of glycosyl azides into glycosyl fluorides.
Carbohydrate research (1993-10-18)
W Bröder, H Kunz
ABSTRACT
Glycosyl azides provide reliable anomeric protection stable to conditions for hydrolytic removal of ester groups, for reductive opening or release of acetalic diol protection, for the introduction of ether-type protection, and for glycosylation processes. The utility of this anomeric protection is further enhanced as glycosyl azides may be converted into glycosyl fluorides, which can be activated for glycosylation reactions. To this end, glycosyl azides have been subjected to 1,3-dipolar cycloaddition with di-tert-butyl acetylenedicarboxylate. On treatment with hydrogen fluoride-pyridine complex the N-glycosyl triazole derivatives directly give glycosyl fluorides.
Sigma-Aldrich
Di-tert-butyl acetylenedicarboxylate, 98%