Merck
  • Effect of bioactive compounds extracted from euphorbious plants on hematological and biochemical parameters of Channa punctatus.
Effect of bioactive compounds extracted from euphorbious plants on hematological and biochemical parameters of Channa punctatus.
Revista do Instituto de Medicina Tropical de Sao Paulo (2011-10-21)
Jaya Shahi, Ajay Singh
ABSTRACT
Channa punctatus was exposed to four different concentrations of Rutin, Taraxerol and Apigenin. Changes in some hematological parameters of Channa punctatus were assessed to determine the influence of these compounds on test fish. Fish were exposed to sublethal concentrations (80% of LC50 of 24h) of these compounds for one week. Control fish were also administered for one week. Thereafter, blood samples were obtained from the control and experimental fish. Blood was assayed for selected hematological parameters (hematocrit, hemoglobin, red blood cell count, white blood cell count total plasma protein and plasma glucose concentration). The derived hematological indices of mean corpuscular hemoglobin concentration (MCHC), mean corpuscular hemoglobin (MCH) and mean corpuscular volume (MCV) were calculated. Sublethal concentrations of these compounds caused a dose dependent decrease in hemoglobin values coupled with a decrease in hematocrit values and red blood cell counts are an obvious indication of anemia. The total white blood cell counts and the differential white blood cell counts were decreased except for the lymphocytes, where there was a slight increase. Plasma protein and glucose were also lower in exposed fish when compared with control. The hematological indices MCH, MCHC, MCV were also lowered. The result from this study reveals high mortality rate and deleterious consequences on the health of fish subjected to acute exposure of Rutin, Taraxerol and Apigenin and therefore, should not be used directly in aquaculture without having the proper knowledge.
Product Description
Supelco
3β-Taraxerol, analytical standard