Merck
  • Controlled synthesis of homopolymers and block copolymers based on 2-(acetoacetoxy)ethyl methacrylate via RAFT radical polymerisation.
Controlled synthesis of homopolymers and block copolymers based on 2-(acetoacetoxy)ethyl methacrylate via RAFT radical polymerisation.
Chemical communications (Cambridge, England) (2003-03-18)
Theodora Krasia, Rémi Soula, Hans G Börner, Helmut Schlaad
ABSTRACT
Reversible addition-fragmentation chain transfer (RAFT) radical polymerisation enables to synthesise well-defined homopolymers and block copolymers based on 2-(acetoacetoxy)ethyl methacrylate, which are capable to strongly coordinate to metals and metal ions.
Sigma-Aldrich
tert-Butyl α-bromoisobutyrate, ≥98.0%