Merck
  • Cloning of ZNF237, a novel member of the MYM gene family that maps to human chromosome 13q11-->q12.
Cloning of ZNF237, a novel member of the MYM gene family that maps to human chromosome 13q11-->q12.
Cytogenetics and cell genetics (2000-07-15)
J Sohal, A Reiter, J M Goldman, N C Cross
ABSTRACT
We have cloned a novel, widely expressed human gene, ZNF237, that shows extensive similarity to the N-terminal region of ZNF198. Two alternatively spliced regions were identified by RT-PCR; the major splice variant is predicted to encode a 383 amino acid protein that contains a single diverged MYM domain. ZNF237 maps to 13q11-->q12, immediately proximal to ZNF198.