Merck
  • Autoreactivity to lipoate and a conjugated form of lipoate in primary biliary cirrhosis.
Autoreactivity to lipoate and a conjugated form of lipoate in primary biliary cirrhosis.
Gastroenterology (2004-01-16)
Sylvaine F A Bruggraber, Patrick S C Leung, Katsushi Amano, Chao Quan, Mark J Kurth, Michael H Nantz, Gordon D Benson, Judy Van de Water, Velimer Luketic, Thomas E Roche, Aftab A Ansari, Ross L Coppel, M Eric Gershwin
ABSTRACT
Although considerable effort has been directed toward the mapping of peptide epitopes by autoantibodies, the role of nonprotein molecules has been less well studied. The immunodominant autoantigen in primary biliary cirrhosis (PBC), E2 components of pyruvate dehydrogenase complexes (PDC-E2), has a lipoate molecule bonded to the domain to which autoantibodies are directed. We examined sera from patients with PBC (n = 105), primary sclerosing cholangitis (n = 70), and rheumatoid arthritis (n = 28) as well as healthy volunteers (n = 43) for reactivity against lipoic acid. The lipoic acid hapten specificity of the reactive antibodies in PBC sera was determined following incubation of aliquots of the sera with human serum albumin (HSA), lipoylated HSA (HSA-LA), PDC-E2, lipoylated PDC-E2, polyethylene glycol (PEG), lipoylated PEG, free lipoic acid, and synthetic molecular mimics of lipoic acid. Anti-lipoic acid specific antibodies were detected in 81% (79 of 97) of antimitochondrial antibody (AMA)-positive patients with PBC but not in controls. Two previously unreported specificities in AMA-positive sera that recognize free lipoic acid and a carrier-conjugated form of lipoic acid were also identified. We hypothesize that conjugated form(s) of native or xenobiotic lipoic acid mimics contribute to the initiation and perpetuation of autoimmunity by at first breaking self-tolerance and participating in subsequent determinant spreading. The variability in the immunoreactive carrier/lipoate conjugates provides an experimental framework on which potential mechanisms for the breakdown of self-tolerance following exposure to xenobiotics can be investigated. The data have implications for patients taking lipoic acid as a dietary supplement.
Sigma-Aldrich
(R)-(+)-α-Lipoic acid, ≥98.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
(R)-(+)-α-Lipoic acid, analytical standard