Merck
  • Cultivating previously uncultured soil bacteria using a soil substrate membrane system.
Cultivating previously uncultured soil bacteria using a soil substrate membrane system.
Nature protocols (2008-08-21)
Belinda C Ferrari, Tristrom Winsley, Michael Gillings, Svend Binnerup
ABSTRACT
Most bacteria are recalcitrant to traditional cultivation in the laboratory. The soil substrate membrane system provides a simulated environment for the cultivation of previously undescribed soil bacteria as microcolonies. The system uses a polycarbonate membrane as a solid support for growth and soil extract as the substrate. Diverse microcolonies can be visualized using total bacterial staining combined with fluorescence in situ hybridization (FISH) after 7-10-d incubation. Molecular typing shows that the majority of microcolony-forming bacteria recovered using this protocol were resistant to growth using standard methods. The protocol takes <4 h of bench time over the 10-d period.
Sigma-Aldrich
Pyruvic acid, 98%