Merck
  • Direct determination of estriol 3- and 16-glucuronides in pregnancy urine by column-switching high-performance liquid chromatography with fluorescence detection.
Direct determination of estriol 3- and 16-glucuronides in pregnancy urine by column-switching high-performance liquid chromatography with fluorescence detection.
Journal of chromatography. B, Biomedical sciences and applications (1997-08-01)
T Iwata, T Hirose, M Yamaguchi
ABSTRACT
An HPLC method for the direct and simultaneous determination of estriol 3- and 16-glucuronides in pregnancy urine is described. The method is based on direct derivatization of the glucuronic acid moiety in estriol glucuronides in urine with 6,7-dimethoxy-1-methyl-2(1H)-quinoxalinone-3-propionylcarboxylic acid hydrazide. The derivatization reaction proceeds in aqueous solution (or urine sample) in the presence of pyridine and 1-ethyl-3-(3-dimethylaminopropyl) carbodiimide at 37 degrees C. The resulting fluorescent derivatives were separated by column-switching chromatography using a first column (YMC-Pack C4) for clean-up of the derivatives and a second column (YMC Pack Ph) for the complete separation of the derivatives. The derivatives were detected spectrofluorimetrically at 445 nm with excitation at 367 nm. The detection limits (signal-to-noise ratio=3) for estriol 3- and 16-glucuronides were 150 and 180 fmol in a 5 microl of urine (14 and 17 ng ml(-1) urine), respectively. The present method is highly sensitive and simple without any clean-up such as conventional solid-phase extraction.
Sigma-Aldrich
Estriol 16α-(β-D-glucuronide), ≥97%