Merck
  • The GABAB receptor antagonist CGP 36,742 and the nootropic oxiracetam facilitate the formation of long-term memory.
The GABAB receptor antagonist CGP 36,742 and the nootropic oxiracetam facilitate the formation of long-term memory.
Behavioural brain research (1996-05-01)
C Mondadori, H J Möbius, J Borkowski
ABSTRACT
The memory-enhancing effects of a single treatment with the GABAB antagonist CGP 36,742 (10 mg/kg) or the nootropic agent oxiracetam (100 mg/kg) given immediately after a learning experience ('post-trial') remain detectable for at least 4 months thereafter. This indicates that in all probability these substances facilitate the formation of the long-term memory trace.