Merck
  • Comparison of the hepatic effects of coumarin, 3,4-dimethylcoumarin, dihydrocoumarin and 6-methylcoumarin in the rat.
Comparison of the hepatic effects of coumarin, 3,4-dimethylcoumarin, dihydrocoumarin and 6-methylcoumarin in the rat.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1994-08-01)
B G Lake, J G Evans, D F Lewis, R J Price
ABSTRACT
The mechanism of coumarin-induced hepatotoxicity in the rat has been investigated by comparing the effects of coumarin with those of three coumarin derivatives, namely 3,4-dihydrocoumarin (DHC), 3,4-dimethylcoumarin (3,4-DMC) and 6-methylcoumarin (6-MC). Male Sprague-Dawley rats were fed either control diet or diets containing 0.5 or 0.75% coumarin, 0.76% DHC, 0.6 or 0.9% 3,4-DMC or 0.82% 6-MC for 13 wk. The dietary levels of 0.5% coumarin and 0.6% 3,4-DMC, were equimolar (3.43 mmol/100 g diet), as were the dietary levels of 0.75% coumarin, 0.76% DHC, 0.9% 3,4-DMC and 0.82% 6-MC (5.14 mmol/100 g diet). All treatments resulted in an increase in relative liver weight, but only coumarin increased plasma alanine aminotransferase and aspartate aminotransferase activities. Morphological examination of liver sections from coumarin treated rats revealed vacuolation of centrilobular hepatocytes and bile duct hyperplasia. Cholangiofibrosis was also observed, particularly in rats given 0.75% coumarin. Treatment with DHC produced no abnormalities, whereas a slight hypertrophy of centrilobular hepatocytes was observed in some 3,4-DMC treated animals and a slight vacuolation of individual hepatocytes was noted in some 6-MC treated rats. DHC, 6-MC and particularly 3,4-DMC treatment resulted in an induction of cytochrome P-450 dependent mixed function oxidase enzyme activities. All treatments induced hepatic GSHS-transferase and gamma-glutamyltransferase activities, induction being most marked in rats given coumarin and 6-MC. These results provide further evidence that coumarin-induced hepatotoxicity in the rat is due to the formation of a 3,4-epoxide intermediate.
Sigma-Aldrich
6-Methylcoumarin, ≥99%, FCC, FG
Sigma-Aldrich
6-Methylcoumarin, ≥99%