Merck
  • Effect of PTSD diagnosis and contingency management procedures on cocaine use in dually cocaine- and opioid-dependent individuals maintained on LAAM: a retrospective analysis.
Effect of PTSD diagnosis and contingency management procedures on cocaine use in dually cocaine- and opioid-dependent individuals maintained on LAAM: a retrospective analysis.
The American journal on addictions (2010-02-19)
Michael J Mancino, Janette McGaugh, Zachary Feldman, James Poling, Alison Oliveto
ABSTRACT
This randomized clinical trial retrospectively examined the effect of post-traumatic stress disorder (PTSD) and contingency management (CM) on cocaine use in opioid and cocaine dependent individuals maintained on high or low-dose LAAM randomly assigned to CM or a yoked-control condition. Cocaine-positive urines decreased more rapidly over time in those without PTSD versus those with PTSD in the noncontingency condition. In participants with PTSD, CM resulted in fewer cocaine-positive urines compared to the noncontingent condition. This suggests that CM may help improve the potentially worse outcomes in opioid- and cocaine-dependent individuals with PTSD compared to those without PTSD. (Am J Addict 2010;00:1-9).