Merck
  • Determination of naltrexone and 6beta-naltrexol in human blood: comparison of high-performance liquid chromatography with spectrophotometric and tandem-mass-spectrometric detection.
Determination of naltrexone and 6beta-naltrexol in human blood: comparison of high-performance liquid chromatography with spectrophotometric and tandem-mass-spectrometric detection.
Analytical and bioanalytical chemistry (2009-12-01)
Sonja Brünen, Ralf Krüger, Susann Finger, Felix Korf, Falk Kiefer, Klaus Wiedemann, Karl J Lackner, Christoph Hiemke
ABSTRACT
We present data for a comparison of a liquid-chromatographic method coupled with tandem mass spectrometry (LC-MS/MS) and a high-performance liquid-chromatographic method with column switching and UV spectrophotometric detection. The two methods were developed for determination of naltrexone and 6beta-naltrexol in blood serum or plasma aiming to be used for therapeutic drug monitoring to guide the treatment of patients with naltrexone. For the high-performance liquid chromatography (HPLC)/UV detection, online sample cleanup was conducted on Perfect Bond C(18) material with 2% (vol/vol) acetonitrile in deionized water. Drugs were separated on a C(18) column using 11.5% (vol/vol) acetonitrile and 0.4% (vol/vol) N,N,N,N-tetramethylethylenediamine within 20 min. LC-MS/MS used naltrexone-d (3) and 6beta-naltrexol-d (4) as internal standards. After protein precipitation, the chromatographic separation was performed on a C(18) column by applying a methanol gradient (5-100%, vol/vol) with 0.1% formic acid over 9.5 min. The HPLC/UV method was found to be linear for concentrations ranging from 2 to 100 ng/ml, with a regression correlation coefficient of r (2) > 0.998 for naltrexone and 6beta-naltrexol. The limit of quantification was 2 ng/ml for naltrexone and 6beta-naltrexol. For the LC-MS/MS method the calibration curves were linear (r(2) > 0.999) from 0.5 to 200 ng/ml for both substances, and the limit of quantification was 0.5 ng/ml. The concentrations measured by the two methods correlated significantly for both substances (r(2) > 0.967; p < 0.001). Both methods could be used for therapeutic drug monitoring. The HPLC/UV method was advantageous regarding automatization and costs, whereas LC-MS/MS was superior with regard to sensitivity.
Product Description
Supelco
6β-Naltrexol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®