Merck
  • Development and validation of chemometrics-assisted spectrophotometric and liquid chromatographic methods for the simultaneous determination of two multicomponent mixtures containing bronchodilator drugs.
Development and validation of chemometrics-assisted spectrophotometric and liquid chromatographic methods for the simultaneous determination of two multicomponent mixtures containing bronchodilator drugs.
Journal of pharmaceutical and biomedical analysis (2006-10-19)
Alaa El-Gindy, Samy Emara, Heba Shaaban
ABSTRACT
Three methods are developed for the determination of two multicomponent mixtures containing guaiphenesine (GU) with salbutamol sulfate (SL), methylparaben (MP) and propylparaben (PP) [mixture 1]; and acephylline piperazine (AC) with bromhexine hydrochloride (BX), methylparaben (MP) and propylparaben (PP) [mixture 2]. The resolution of the two multicomponent mixtures has been accomplished by using numerical spectrophotometric methods such as partial least squares (PLS-1) and principal component regression (PCR) applied to UV absorption spectra of the two mixtures. In addition HPLC method was developed using a RP 18 column at ambient temperature with mobile phase consisting of acetonitrile-0.05 M potassium dihydrogen phosphate, pH 4.3 (60:40, v/v), with UV detection at 243 nm for mixture 1, and mobile phase consisting of acetonitrile-0.05 M potassium dihydrogen phosphate, pH 3 (50:50, v/v), with UV detection at 245 nm for mixture 2. The methods were validated in terms of accuracy, specificity, precision and linearity in the range of 20-60 microg ml(-1) for GU, 1-3 microg ml(-1) for SL, 20-80 microg ml(-1) for AC, 0.2-1.8 microgml(-1) for PP and 1-5 microg ml(-1) for BX and MP. The proposed methods were successfully applied for the determination of the two multicomponent combinations in laboratory prepared mixtures and commercial syrups.