Merck
  • Investigation of the agonist activity of prostacyclin analogues on prostanoid EP4 receptors using GW 627368 and taprostene: evidence for species differences.
Investigation of the agonist activity of prostacyclin analogues on prostanoid EP4 receptors using GW 627368 and taprostene: evidence for species differences.
Prostaglandins, leukotrienes, and essential fatty acids (2005-03-15)
R L Jones, K M Chan
ABSTRACT
The possibility that the prostacyclin analogues AFP-07 and cicaprost relax saphenous vein preparations of pig, guinea-pig and rabbit by simultaneously activating prostanoid EP4 and IP (prostacyclin) receptors was investigated using the high-affinity EP4 antagonist GW 627368. The IP receptor system in each preparation was suppressed with the partial agonist taprostene. The results indicate that AFP-07 and cicaprost are moderately potent EP4 agonists on pig saphenous vein, but much weaker EP4 agonists on guinea-pig saphenous vein. GW 627368 did not antagonise AFP-07- and cicaprost-induced relaxation of rabbit saphenous vein, which contrasts with a previous study using the EP4 blocker AH 23848. However, treatment with taprostene was of less value due to poorer antagonism of the rabbit IP system; this may be related to the presence of a sensitive EP2 relaxant system. Relaxation of each preparation induced by the selective EP2 agonist ONO-AE1-259 was unaffected by GW 627368, with and without taprostene.