Merck
  • Preliminary evaluation of anticonvulsant activity of some 4-(benzyloxy)-benzamides.
Preliminary evaluation of anticonvulsant activity of some 4-(benzyloxy)-benzamides.
Acta poloniae pharmaceutica (2004-04-15)
Henryk Marona, Edward Szneler
ABSTRACT
A series of alkanolamides have been tested for anticonvulsant activity in the maximal electroshock seizure (MES) and subcutaneous pentylenetetrazole seizure treshold (ScMet) assays and for neurotoxicity (TOX) in rodents. Most interesting were the anticonvulsant results of 2N-methylaminoethanol derivative II, which displayed anti-MES activity (mice) with a protective index (TD50/ED50) of 2.536 higher than that for valproate.
Sigma-Aldrich
4-Benzyloxybenzoic acid, 99%