Merck
Progressive supranuclear palsy syndrome induced by clebopride.
Movement disorders : official journal of the Movement Disorder Society (2004-04-13)
Jaume Campdelacreu, Hatice Kumru, Eduard Tolosa, Josep Valls-Solé, Antoni Benabarre
ABSTRACT
We report on a patient who presented with a progressive supranuclear palsy (PSP) syndrome while receiving clebopride (CLB), a prokinetic drug with central antidopaminergic properties. The clinical and neurophysiological signs progressively disappeared after CLB withdrawal. To our knowledge, this is the first published PSP-like syndrome attributable to an antidopaminergic drug.