- Bioactivation and cytotoxicity of 1,1-dichloro-2,2,2-trifluoroethane (HCFC-123) in isolated rat hepatocytes.
Bioactivation and cytotoxicity of 1,1-dichloro-2,2,2-trifluoroethane (HCFC-123) in isolated rat hepatocytes.
The bioactivation and cytotoxicity of 1,1-dichloro-2,2,2-trifluoroethane (HCFC-123), a replacement for some ozone-depleting chlorofluorocarbons, were investigated using freshly isolated hepatocytes from non-induced male rats. A time- and concentration-dependent increase in the leakage of lactate dehydrogenase and a concentration-dependent loss of total cellular glutathione were observed in cells incubated with 1, 5 and 10 mM HCFC-123 under normoxic or hypoxic (about 4% O2) conditions. Lactate dehydrogenase leakage was completely prevented by pretreating the cell suspension with the free radical trapper N-t-butyl-alpha-phenylnitrone. The aspecific cytochrome P450 (P450) inhibitor, metyrapone, totally prevented the lactate dehydrogenase leakage from hepatocytes, while two isoform-specific P450 inhibitors, 4-methylpyrazole and troleandomycin (a P450 2E1 and a P450 3A inhibitor, respectively), provided a partial protection against HCFC-123 cytotoxicity. Interestingly, pretreatment of cells with glutathione depletors, such as phorone and diethylmaleate, did not enhance the HCFC-123-dependent lactate dehydrogenase leakage. Two stable metabolites of HCFC-123, 1-chloro-2,2,2-trifluoroethane and 1-chloro-2,2-difluoroethene, were detected by gas chromatography/mass spectrometry analysis of the head space of the hepatocyte incubations carried out under hypoxic and, although at a lower level, also normoxic conditions, indicating that reductive metabolism of HCFC-123 by hepatocytes had occurred. The results overall indicate that HCFC-123 is cytotoxic to rat hepatocytes under both normoxic and hypoxic conditions, due to its bioactivation to reactive metabolites, probably free radicals, and that P450 2E1 and, to a lower extent, P450 3A, are involved in the process.