Merck
  • Contact allergy to 3-methylol phenol, 2,4-dimethylol phenol and 2,6-dimethylol phenol.
Contact allergy to 3-methylol phenol, 2,4-dimethylol phenol and 2,6-dimethylol phenol.
Acta dermato-venereologica (1985-01-01)
M Bruze, E Zimerson
ABSTRACT
Thirteen patients with contact allergy to phenol-formaldehyde resins (P-F-R) were patch tested with 3-methylol phenol, 2,4-dimethylol phenol and 2,6-dimethylol phenol. Nine patients reacted to at least 1 compound, all giving positive test responses to 2,4-dimethylol phenol. Seven patients reacted simultaneously to 2,6-dimethylol phenol while only 1 patient reacted to 3-methylol phenol. Negative test responses were noted in 20 controls. Chemical investigation by high pressure liquid chromatography indicated that the compounds tested were pure and separable. The 3 reported sensitizers may, theoretically, be generated during the manufacture of P-F-R. 2,4-Dimethylol phenol and 2,6-dimethylol phenol have been demonstrated and there has been chromatographic evidence of 3-methylol phenol in the P-F-R used in the routine test series at the department.
Sigma-Aldrich
3-Hydroxybenzyl alcohol, 99%