Merck
  • Estradiol 17 beta-D-glucuronide is a high-affinity substrate for oatp organic anion transporter.
Estradiol 17 beta-D-glucuronide is a high-affinity substrate for oatp organic anion transporter.
The American journal of physiology (1996-02-01)
N Kanai, R Lu, Y Bao, A W Wolkoff, M Vore, V L Schuster
ABSTRACT
Although substantial evidence indicates that estradiol-17 beta (E2) is conjugated to the glucuronide in the kidney and then excreted by a direct tubular secretory route and that the liver transports E2 glucuronides via carrier-mediated mechanisms, the transporters involved in these processes have not been identified. The so-called "organic anion-transporting polypeptide" (i.e., oatp) has a number of known substrates, including bromosulfophthalein (BSP) and taurocholic acid (TCA) (E. Jacquemin, B. Hagenbuch, B. Stieger, A. W. Wolkoff, and P. J. Meier. Proc. Natl. Acad. Sci. USA 91: 133-137, 1994). In a companion study, we determined that steroid hormones represent a class of hormones that interact strongly with oatp when the latter is transiently expressed in vitro. Here, we studied more extensively steroids and steroid anion conjugates as candidate oatp substrates. In HeLa cell monolayers transfected with a full-length oatp cDNA, [3H]estradiol 17 beta-D-glucuronide ([3H]E2-17G) was transported with a signal-to-noise ratio of 15:1 over that of monolayers transfected with a control plasmid. The affinity of oatp for [3H]E2-17G was significantly higher than that for TCA (K(m) of 3 microM vs. 27 microM, respectively). In contrast to E2-17G, unconjugated estradiol (E2) was not significantly transported by oatp. Several unconjugated steroids and anionic steroid conjugates were tested for their ability to compete with tracer E2-17G for oatp-mediated transport. Conjugation at the 17 or 3 position with the anion of a strong acid (sulfate) resulted in a greater degree of inhibition of tracer E2-17G transport than did conjugation at the 17 or 3 position with an uncharged group (acetate), suggesting that the strength of the negative charge at these positions is an important determinant of the affinity of a given steroid conjugate for oatp. We conclude that the preferred substrates for oatp are steroids with a strong 17- or 3-position anionic group. Since steroid sulfotransferases and glucuronosyltransferases are expressed in the proximal tubule, as is oatp, the transporter may serve as an apical exit pathway for steroids following their conjugation within the tubule cell.
Sigma-Aldrich
β-Estradiol 3-(β-D-glucuronide) sodium salt