Merck
Thiodicarb.
Acta crystallographica. Section C, Crystal structure communications (1995-08-15)
H L Ammon, R J Seymour, W M Huffman, T D D'Silva
ABSTRACT
Dimethyl N,N'-[thiobis(methyliminocarbonyloxy)] bis-(ethanimidothiolate), C10H18N4O4S3, is an example of a sulfenylated biscarbamate insecticide. The molecule has an approximate twofold axis through the central S atom which joins the two methyliminocarbonyloxyethanimidothiolate units. One of the two arms is planar in the crystal. Semi-empirical geometry optimization for an isolated molecule favors a model with both arms planar and, thus, crystal packing may be responsible for the observed non-planarity in one arm. Bond lengths and angles have similar values to those of the 'monomeric' carbamate insecticide methomyl.
Product Description
Supelco
Thiodicarb, PESTANAL®, analytical standard