Merck
  • Heat inactivation of paraoxonase and arylesterase activities in human and rabbit serum.
Heat inactivation of paraoxonase and arylesterase activities in human and rabbit serum.
Chemico-biological interactions (1993-06-01)
V Simeon, E Pavković
ABSTRACT
The heat inactivation of esterases in human and rabbit serum was followed at 50 and 55 degrees C by measuring the decrease of activity with paraoxon, phenylacetate and beta-naphthylacetate as substrates. The rate of inactivation measured with the three substrates was slightly, but significantly different, indicating that the substrates are hydrolysed by different enzymes.
Sigma-Aldrich
2-Naphthaleneacetic acid, 99%