Merck
  • [Method for determining the age of the fibrin in the disseminated intravascular coagulation syndrome].
[Method for determining the age of the fibrin in the disseminated intravascular coagulation syndrome].
Arkhiv patologii (1984-01-01)
D D Zerbino, L L Lukasevich
ABSTRACT
Three methods for the detection of generalized intravascular coagulation are proposed. The dyes produced in the USSR are used in these methods: acid red 2C--orange-red-blue method; acid yellow light-proof--picric red-blue method; acid blue-black in the method called red-blue-black. The use of these methods allows one to determine the duration of the fibrin existence in the vessels and to distinguish "young", "mature" and "old" fibrin.
Product Description
Supelco
Yellow 2G, analytical standard
Sigma-Aldrich
Acid Yellow 17, Dye content 60 %