Merck
  • Response profiles of enteric methane emissions and lactational performance during habituation to dietary coconut oil in dairy cows.
Response profiles of enteric methane emissions and lactational performance during habituation to dietary coconut oil in dairy cows.
Journal of dairy science (2013-01-15)
M Hollmann, W J Powers, A C Fogiel, J S Liesman, D K Beede
ABSTRACT
Dietary coconut oil (CNO) can reduce dry matter intake (DMI), enteric methane (eCH(4)) emissions, and milk fat yield of lactating cows. The goals of this research were to examine responses to different CNO concentrations during the habituation period (34-d) and to evaluate temporal patterns of DMI, eCH(4), and milk fat yield. Treatment diets contained (dry basis): 0.0% (CNO0), 1.3% (CNO1.3), 2.7% (CNO2.7), 3.3% (CNO3.3), or 4.0% CNO (CNO4). In experiment 1, 12 primi- or small secundiparous cows were housed in individual, environmentally controlled rooms and fed CNO0, CNO1.3, CNO2.7, or CNO4. Measurements included DMI, eCH(4), and milk yield and composition. Due to a precipitous drop in DMI (26%), cows fed CNO4 were replaced with cows fed CNO3.3 following d 10. Dietary CNO of 2.7% or more reduced eCH(4) emissions. Reduction was greater with increased CNO and during the first than the second half of the day. Simultaneously, decline in DMI of cows fed CNO2.7, CNO3.3, or CNO4 was increasingly precipitous with increased CNO concentration. Total-tract neutral detergent fiber (NDF) digestibility during wk 5 was reduced in cows fed CNO2.7 or CNO3.3, which in part explained concomitantly reduced eCH(4)/DMI. In addition, milk fat yield was depressed at an increasing rate in cows fed CNO2.7, CNO3.3, and CNO4. In experiment 2, DMI was measured individually in 12 multiparous cows during habituation to CNO0, CNO1.3, CNO2.7, or CNO3.3 for 21 d before relocation to individual, environmentally controlled rooms. Dietary CNO2.7 or CNO3.3 reduced DMI by d 4 and total-tract NDF digestibility during wk 5. Relocation to individual rooms was associated with a 15% reduction in DMI, which was not affected by treatment. Results showed that 2.7% or more dietary CNO reduced eCH(4) and DMI, caused milk fat depression, and decreased NDF digestibility.
Sigma-Aldrich
Coconut oil, virgin, Certified organic (NOP)
Sigma-Aldrich
Coconut oil from Cocos nucifera, low-melting solid