Merck
  • Acipimox attenuates atherosclerosis and enhances plaque stability in ApoE-deficient mice fed a palmitate-rich diet.
Acipimox attenuates atherosclerosis and enhances plaque stability in ApoE-deficient mice fed a palmitate-rich diet.
Biochemical and biophysical research communications (2012-10-13)
Feipeng Jin, Sihua Jiang, Dachun Yang, Xingmei Zhang, Yun Yang, Yan Zhang, Kun Li, Yongjian Yang, Shuangtao Ma
ABSTRACT
Saturated fatty acids (FA) have been linked to an increased risk of cardiovascular disease. The effects of acipimox, a FA-lowering agent, on palmitate- (an important saturated fatty acid) stimulated atherosclerosis remains to be elucidated. We investigated the effects of acipimox on atherosclerosis in ApoE(-/-) mice fed a palmitate-rich diet. Male ApoE(-/-) mice, 6-8 weeks of age, were randomized into three groups. The animals were fed a normal chow diet in the control group, a diet containing 5% palmitic acid in the palmitate group, and a diet containing 5% palmitic acid and 0.02% acipimox in the acipimox group. The plasma lipid profiles, aortic lesions, plaque collagen content and the expression of matrix metalloproteinase (MMP)-2, MMP-3, MMP-9, and MMP-14 and the tissue inhibitor of MMP (TIMP)-1, and TIMP-2 were determined after a 12-week treatment. The palmitate-rich diet significantly increased plasma FA concentrations (P<0.01), enhanced atherosclerotic lesions (P<0.01), decreased plaque collagen content (P<0.01) and upregulated MMP-2 (P<0.05) in the aorta. Additionally, all of these harmful effects were significantly attenuated by co-treatment with acipimox (P<0.05 or P<0.01). The present study suggests that acipimox attenuates atherosclerosis and enhances plaque stability in ApoE(-/-) mice fed a palmitate-rich diet.
Sigma-Aldrich
Acipimox, ≥99% (TLC)