Merck
Analysis of a homemade Edison tinfoil phonograph.
The Journal of the Acoustical Society of America (2012-10-09)
Jason D Sagers, Andrew R McNeese, Richard D Lenhart, Preston S Wilson
ABSTRACT
Thomas Edison's phonograph was a landmark acoustic invention. In this paper, the phonograph is presented as a tool for education in acoustics. A brief history of the phonograph is outlined and an analogous circuit model that describes its dynamic response is discussed. Microphone and scanning laser Doppler vibrometer (SLDV) measurements were made on a homemade phonograph for model validation and inversion for unknown model parameters. SLDV measurements also conclusively illustrate where model assumptions are violated. The model elements which dominate the dynamic response are discussed.
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis