Merck
  • A survey of aflatoxin M1 contamination in bulk milk samples from dairy bovine, ovine, and caprine herds in Iran.
A survey of aflatoxin M1 contamination in bulk milk samples from dairy bovine, ovine, and caprine herds in Iran.
Bulletin of environmental contamination and toxicology (2012-04-25)
Ebrahim Rahimi, Mehrdad Ameri
ABSTRACT
A total of 150 bovine (60), ovine (42), and caprine (48) bulk milk samples were analyzed using a commercially available competitive ELISA kit. Overall, AFM1 was found in 46.7 % of the analyzed samples by an average concentration of 40.3 ± 22.2 ng/L. The incidence rates of AFM1 contamination in bovine, ovine, and caprine bulk milk samples were 66.7, 31.0, and 35.4 %, respectively. The concentration of AFM1 in 37.5 % of AFM1-positive bovine milk samples and 5.9 % of AFM1-positive caprine milk samples were higher than 50 ng/L.
Sigma-Aldrich
Aflatoxin M1, from Aspergillus flavus
Supelco
Aflatoxin M1 solution, 0.5 μg/mL in acetonitrile, analytical standard