Merck
  • Improved syntheses of 5'-S-(2-aminoethyl)-6-N-(4-nitrobenzyl)-5'-thioadenosine (SAENTA), analogues, and fluorescent probe conjugates: analysis of cell-surface human equilibrative nucleoside transporter 1 (hENT1) levels for prediction of the antitumor efficacy of gemcitabine.
Improved syntheses of 5'-S-(2-aminoethyl)-6-N-(4-nitrobenzyl)-5'-thioadenosine (SAENTA), analogues, and fluorescent probe conjugates: analysis of cell-surface human equilibrative nucleoside transporter 1 (hENT1) levels for prediction of the antitumor efficacy of gemcitabine.
Journal of medicinal chemistry (2010-08-20)
Morris J Robins, Yunshan Peng, Vijaya L Damaraju, Delores Mowles, Geraldine Barron, Tracey Tackaberry, James D Young, Carol E Cass
ABSTRACT
5'-S-(2-aminoethyl)-6-N-(4-nitrobenzyl)-5'-thioadenosine (SAENTA), 5'-S-(2-acetamidoethyl)-6-N-[(4-substituted)benzyl]-5'-thioadenosine analogues, 5'-S-[2-(6-aminohexanamido)]ethyl-6-N-(4-nitrobenzyl)-5'-thioadenosine (SAHENTA), and related compounds were synthesized by S(N)Ar displacement of fluoride from 6-fluoropurine intermediates with 4-(substituted)benzylamines. Conjugation of the pendant amino groups of SAENTA and SAHENTA with fluorescein-5-yl isothiocyanate (FITC) gave fluorescent probes that bound at nanomolar concentrations specifically to human equilibrative nucleoside transporter 1 (hENT1) produced in recombinant form in model expression systems and in native form in cancer cell lines. Transporter binding effects were studied and the ability of the probes to predict the potential antitumor efficacy of 2'-deoxy-2',2'-difluorocytidine (gemcitabine) was demonstrated.
Sigma-Aldrich
S-(4-Nitrobenzyl)-6-thioinosine, ≥98%, solid