Merck
  • Fast and green microwave-assisted conversion of essential oil allylbenzenes into the corresponding aldehydes via alkene isomerization and subsequent potassium permanganate promoted oxidative alkene group cleavage.
Fast and green microwave-assisted conversion of essential oil allylbenzenes into the corresponding aldehydes via alkene isomerization and subsequent potassium permanganate promoted oxidative alkene group cleavage.
Molecules (Basel, Switzerland) (2009-09-29)
Thi X Thi Luu, Trinh To Lam, Thach Ngoc Le, Fritz Duus
ABSTRACT
Essential oil allylbenzenes from have been converted quickly and efficiently into the corresponding benzaldehydes in good yields by a two-step "green" reaction pathway based on a solventless alkene group isomerization by KF/Al(2)O(3) to form the corresponding 1-arylpropene and a subsequent solventless oxidation of the latter to the corresponding benzaldehyde by KMnO(4)/CuSO(4).5H(2)O. The assistance by microwave irradiation results in very short reaction times (<15 minutes). The green conversion of eugenol (4-allyl-2-methoxyphenol) into vanillin (4-hydroxy-3-methoxybenzaldehyde) has been carried out in a similar way, requiring however two additional microwave-assisted synthetic steps for acetylation of the hydroxy group prior to the oxidation reaction, and for the final deacetylation of vanillin acetate (4-acetoxy-3-methoxybenzaldehyde) by KF/Al(2)O(3) under solvent-free conditions, respectively.
Sigma-Aldrich
Allylbenzene, 98%