Merck
  • A MOFRL family glycerate kinase from the thermophilic crenarchaeon, Sulfolobus tokodaii, with unique enzymatic properties.
A MOFRL family glycerate kinase from the thermophilic crenarchaeon, Sulfolobus tokodaii, with unique enzymatic properties.
Biotechnology letters (2009-08-20)
Bo Liu, Lei Wu, Tianming Liu, Ye Hong, Yulong Shen, Jinfeng Ni
ABSTRACT
A glycerate kinase gene (ST2037) from the hyperthermophilic crenarchaeon Sulfolobus tokodaii was cloned and expressed in Escherichia coli. The purified homodimeric protein (45 kDa) specifically catalyzed the formation of 2-phosphoglycerate with D-glycerate as substrate. The thermostable enzyme displayed maximum activity (over 20 min) at 90 degrees C and pH 4.5. The maximal activity was in the presence of Co(2+). The MOFRL family glycerate kinase used AMP as phosphate donor with maximal activity towards GTP. These characteristics of the enzyme suggested its potential in the catalytic production of 2-phosphoglycerate.
Sigma-Aldrich
L-2-Phosphoglyceric acid disodium salt hydrate, ≥80% (CE)